accessibility menu, dialog, popup

UserName

FeP(R)NCH3H3CCH3PhPh

CAS RN 55700-44-2

IUPAC Name: (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
Synonyms: —
Molecular Weight (g/mol): 441.34
Molecular Formula: C26H28FeNP
InChi Key: RCAFPSZHKFOLEE-KVEBOLLKNA-N
SMILES: [Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results