missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific™

Chemical Identifiers

CAS 616867-28-8
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.43
InChI Key AWEZXIRZNQCCNN-OAQYLSRUSA-N
Synonym r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
PubChem CID 11210384
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid
SMILES CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441145000
SDS View Documents
Thermo Scientific Chemicals
441145000
500mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

616867-28-8
353.43
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid
C21H23NO4
AWEZXIRZNQCCNN-OAQYLSRUSA-N
11210384
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

616867-28-8,1634-04-4
100.0
Authentic
C21H23NO4
-8.6
10% max. (MTBE)
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
353.43
353.43
(R)-N-FMOC-α-Methylvaline, 98% ee
97.5
98.0% min.
97.5% min. (HPLC)
500mg
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
AWEZXIRZNQCCNN-OAQYLSRUSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid
11210384
98%,98% ee
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
In case of fire: Use carbon dioxide for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only