missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, Thermo Scientific™
Chemical Identifiers
| CAS | 193086-74-7 |
|---|---|
| Molecular Formula | C25H22FNO4 |
| Molecular Weight (g/mol) | 419.46 |
| InChI Key | VNYGHKCAXQKZEQ-VWLOTQADSA-N |
| Synonym | s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine, fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine |
| PubChem CID | 44598473 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid |
| SMILES | CC(CC1=CC=CC=C1F)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC441115000
|
N/A | 500mg | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 193086-74-7 | |
| 419.46 | |
| s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine, fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid |
| C25H22FNO4 | |
| VNYGHKCAXQKZEQ-VWLOTQADSA-N | |
| 44598473 | |
| CC(CC1=CC=CC=C1F)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Specifications
| 193086-74-7 | |
| 100.0 | |
| Authentic | |
| C25H22FNO4 | |
| s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine, fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine | |
| CC(CC1=CC=CC=C1F)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
| 419.46 | |
| 419.46 | |
| (R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98% ee |
| 97.5 | |
| 98.0% min. | |
| 97.5% min. (HPLC) | |
| 48 | |
| VNYGHKCAXQKZEQ-VWLOTQADSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid | |
| 44598473 | |
| 98%,98% ee |
RUO – Research Use Only