missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-alpha-Ethylalanine, 98%, 98% ee

Catalog No. AC441061000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441061000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441061000 Supplier Thermo Scientific Chemicals Supplier No. 441061000

CAS: 123254-58-0 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123254-58-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD12963786
InChI Key SHZXLTCEPXVCSV-SNVBAGLBSA-N
Synonym boc-d-isovaline, r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, r-2-tert-butoxycarbonylamino-2-methylbutanoic acid, boc-iso-valine, r-n-boc-a-ethylalanine, s-n-boc-a-ethylalanine, r-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2r-2-tert-butoxycarbonylamino-2-methyl-butanoic acid, 2r-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
PubChem CID 14284791
IUPAC Name (2R)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C)(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 104°C to 112°C
CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation -12
Quantity 100 mg
Formula Weight 217.27
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-BOC-α-Ethylalanine, 98% ee
Show More Show Less

RUO – Research Use Only