missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-alpha-Ethylalanine, 98%, 98% ee

CAS: 123254-58-0 | C10H19NO4 | 217.27 g/mol

$266.12 - $266.12

Chemical Identifiers

CAS 123254-58-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD12963786
InChI Key SHZXLTCEPXVCSV-SNVBAGLBSA-N
Synonym boc-d-isovaline, r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, r-2-tert-butoxycarbonylamino-2-methylbutanoic acid, boc-iso-valine, r-n-boc-a-ethylalanine, s-n-boc-a-ethylalanine, r-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2r-2-tert-butoxycarbonylamino-2-methyl-butanoic acid, 2r-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
PubChem CID 14284791
IUPAC Name (2R)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C)(NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441061000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
441061000
100 mg
Each for $266.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123254-58-0
217.27
SHZXLTCEPXVCSV-SNVBAGLBSA-N
14284791
CCC(C)(NC(=O)OC(C)(C)C)C(O)=O
C10H19NO4
MFCD12963786
boc-d-isovaline, r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, r-2-tert-butoxycarbonylamino-2-methylbutanoic acid, boc-iso-valine, r-n-boc-a-ethylalanine, s-n-boc-a-ethylalanine, r-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2r-2-tert-butoxycarbonylamino-2-methyl-butanoic acid, 2r-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
(2R)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

104°C to 112°C
97.5
White to Yellow
98%
Glass Bottle
-12
SHZXLTCEPXVCSV-SNVBAGLBSA-N
(2R)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
217.27
217.27
Powder
123254-58-0
100.0
Authentic
C10H19NO4
MFCD12963786
boc-d-isovaline, r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, r-2-tert-butoxycarbonylamino-2-methylbutanoic acid, boc-iso-valine, r-n-boc-a-ethylalanine, s-n-boc-a-ethylalanine, r-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2r-2-tert-butoxycarbonylamino-2-methyl-butanoic acid, 2r-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
CCC(C)(NC(=O)OC(C)(C)C)C(O)=O
100 mg
14284791
98%,98% ee
(R)-N-BOC-α-Ethylalanine, 98% ee
Videos
SDS
Documents

Documents

RUO – Research Use Only