missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-Benzyl-1-phenylethylamine 98.0+%, TCI America™

$61.51 - $175.22

Chemical Identifiers

CAS 38235-77-7
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD00015010
InChI Key ZYZHMSJNPCYUTB-CYBMUJFWSA-N
Synonym r-+-n-benzyl-1-phenylethylamine, r-n-benzyl-1-phenylethanamine, r-+-n-benzyl-alpha-methylbenzylamine, benzyl 1r-1-phenylethyl amine, r-n-benzyl-1-phenylethylamine, 1r-1-phenylethyl benzylamine, 1r-n-benzyl-1-phenylethanamine, s---n-benzyl-alpha-phenethylamine, pubchem5974
PubChem CID 1268086
IUPAC Name (1R)-N-benzyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)NCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B15285ML
View Documents
TCI America
B15285ML
5 mL N/A
Only null left
 
B152825ML
View Documents
TCI America
B152825ML
25 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

38235-77-7
211.308
ZYZHMSJNPCYUTB-CYBMUJFWSA-N
1268086
CC(C1=CC=CC=C1)NCC2=CC=CC=C2
C15H17N
MFCD00015010
r-+-n-benzyl-1-phenylethylamine, r-n-benzyl-1-phenylethanamine, r-+-n-benzyl-alpha-methylbenzylamine, benzyl 1r-1-phenylethyl amine, r-n-benzyl-1-phenylethylamine, 1r-1-phenylethyl benzylamine, 1r-n-benzyl-1-phenylethanamine, s---n-benzyl-alpha-phenethylamine, pubchem5974
(1R)-N-benzyl-1-phenylethanamine

Specifications

38235-77-7
C15H17N
5 mL
ZYZHMSJNPCYUTB-CYBMUJFWSA-N
(1R)-N-benzyl-1-phenylethanamine
1268086
≥98.0% (GC)
(R)-(+)-N-Benzyl-1-phenylethylamine
Yellow
MFCD00015010
r-+-n-benzyl-1-phenylethylamine, r-n-benzyl-1-phenylethanamine, r-+-n-benzyl-alpha-methylbenzylamine, benzyl 1r-1-phenylethyl amine, r-n-benzyl-1-phenylethylamine, 1r-1-phenylethyl benzylamine, 1r-n-benzyl-1-phenylethanamine, s---n-benzyl-alpha-phenethylamine, pubchem5974
CC(C1=CC=CC=C1)NCC2=CC=CC=C2
211.308
211.31
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'