missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-Benzyl-1-phenylethylamine 98.0+%, TCI America™

Catalog No. B15285ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15285ML 5 mL
B152825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15285ML Supplier TCI America Supplier No. B15285ML
Only null left

Chemical Identifiers

CAS 38235-77-7
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD00015010
InChI Key ZYZHMSJNPCYUTB-CYBMUJFWSA-N
Synonym r-+-n-benzyl-1-phenylethylamine, r-n-benzyl-1-phenylethanamine, r-+-n-benzyl-alpha-methylbenzylamine, benzyl 1r-1-phenylethyl amine, r-n-benzyl-1-phenylethylamine, 1r-1-phenylethyl benzylamine, 1r-n-benzyl-1-phenylethanamine, s---n-benzyl-alpha-phenethylamine, pubchem5974
PubChem CID 1268086
IUPAC Name (1R)-N-benzyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)NCC2=CC=CC=C2

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 211.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-N-Benzyl-1-phenylethylamine