missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-Methylsuccinic Acid 98.0+%, TCI America™
$214.37 - $741.89
Chemical Identifiers
| CAS | 3641-51-8 |
|---|---|
| Molecular Formula | C5H8O4 |
| Molecular Weight (g/mol) | 132.12 |
| MDL Number | MFCD00209602 |
| InChI Key | WXUAQHNMJWJLTG-GSVOUGTGSA-N |
| Synonym | (R)-(+)-Pyrotartaric Acid |
| PubChem CID | 77983 |
| IUPAC Name | (2R)-2-methylbutanedioic acid |
| SMILES | C[C@H](CC(O)=O)C(O)=O |
Chemical Identifiers
| 3641-51-8 | |
| 132.12 | |
| WXUAQHNMJWJLTG-GSVOUGTGSA-N | |
| 77983 | |
| C[C@H](CC(O)=O)C(O)=O |
| C5H8O4 | |
| MFCD00209602 | |
| (R)-(+)-Pyrotartaric Acid | |
| (2R)-2-methylbutanedioic acid |
Specifications
| 3641-51-8 | |
| Purple-White | |
| MFCD00209602 | |
| (R)-(+)-Pyrotartaric Acid | |
| C[C@H](CC(O)=O)C(O)=O | |
| 132.12 | |
| 132.12 | |
| Crystalline Powder |
| 114°C | |
| C5H8O4 | |
| 5 g | |
| WXUAQHNMJWJLTG-GSVOUGTGSA-N | |
| (2R)-2-methylbutanedioic acid | |
| 77983 | |
| ≥98.0% (GC,T) | |
| (R)-(+)-Methylsuccinic Acid |
Safety and Handling
EINECSNumber : (2)-0903
TSCA : No