missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Methylsuccinic Acid 98.0+%, TCI America™

Catalog No. M122625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M122625G 25 g
M12265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M122625G Supplier TCI America Supplier No. M122625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3641-51-8
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00209602
InChI Key WXUAQHNMJWJLTG-GSVOUGTGSA-N
Synonym (R)-(+)-Pyrotartaric Acid
PubChem CID 77983
IUPAC Name (2R)-2-methylbutanedioic acid
SMILES C[C@H](CC(O)=O)C(O)=O

Specifications

Melting Point 114°C
Color Purple-White
Quantity 25 g
Formula Weight 132.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-Methylsuccinic Acid
EINECSNumber (2)-0903
TSCA No