missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methoxyphenylacetic acid, 99%

Catalog No. AC224470010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224470010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC224470010 Supplier Thermo Scientific Chemicals Supplier No. 224470010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3966-32-3 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent

Chemical Identifiers

CAS 3966-32-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004250
InChI Key DIWVBIXQCNRCFE-MRVPVSSYSA-N
Synonym r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j
PubChem CID 2724294
IUPAC Name (2R)-2-methoxy-2-phenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 67°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH(OCH3)COOH
Quantity 1 g
Beilstein 10, 195
Specific Rotation Condition −160.40° (20°C c=1,methanol)
Specific Rotation -160.4
Formula Weight 166.18
Percent Purity 99%
Physical Form Fine Crystalline Powder or Needles
Chemical Name or Material (R)-(-)-α-Methoxyphenylacetic acid
Show More Show Less
EINECSNumber 223-580-9

RUO – Research Use Only