missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-α-Lipoic acid, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012741
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012741 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012741 Supplier Merck Emd Millipore Supplier No. 04471100MG
Only null left

α-Lipoic acid is an antioxidant which is generally present in plants, animals and many other microorganisms.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 1200-22-2
Flash Point >110°C
Linear Formula C8H14O2S2
MDL Number MFCD01631142
Melting Point 48°C to 52°C
Synonym (R)-(+)-1,2-Dithiolane-3-pentanoic acid; (R)-6,8-Dithiooctanoic acid; (R)-6,8-Thioctic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 206.33
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less