missing translation for 'onlineSavingsMsg'
Learn More

(R)-Epichlorohydrin 98.0+%, TCI America™

Supplier:  TCI America E058125G

 View more versions of this product

Catalog No. E058125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(R)-Epichlorohydrin
Yellow
C3H5ClO
25 g
r---epichlorohydrin, r-epichlorohydrin, 2r-2-chloromethyl oxirane, --epichlorohydrin, r-chloromethyl oxirane, r-2-chloromethyl oxirane, epichlorohydrin,-, unii-vec01h609i, r-1-chloro-2,3-epoxypropane
ClC[C@H]1CO1
92.52
CHEBI:18662
≥98.0% (GC)
51594-55-9
116°C
MFCD00077759
2023
BRLQWZUYTZBJKN-VKHMYHEASA-N
(2R)-2-(chloromethyl)oxirane
2734062
92.52
Liquid

Chemical Identifiers

51594-55-9
92.52
BRLQWZUYTZBJKN-VKHMYHEASA-N
2734062
(2R)-2-(chloromethyl)oxirane
C3H5ClO
MFCD00077759
r---epichlorohydrin, r-epichlorohydrin, 2r-2-chloromethyl oxirane, --epichlorohydrin, r-chloromethyl oxirane, r-2-chloromethyl oxirane, epichlorohydrin,-, unii-vec01h609i, r-1-chloro-2,3-epoxypropane
CHEBI:18662
ClC[C@H]1CO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0275

RTECSNumber : RR0427000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions