missing translation for 'onlineSavingsMsg'
Learn More

(R)-Epichlorohydrin 98.0+%, TCI America™

Catalog No. p-7129076
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05815G 5 g
E058125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05815G Supplier TCI America Supplier No. E05815G

Chemical Identifiers

CAS 51594-55-9
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00077759
InChI Key BRLQWZUYTZBJKN-VKHMYHEASA-N
Synonym r---epichlorohydrin, r-epichlorohydrin, 2r-2-chloromethyl oxirane, --epichlorohydrin, r-chloromethyl oxirane, r-2-chloromethyl oxirane, epichlorohydrin,-, unii-vec01h609i, r-1-chloro-2,3-epoxypropane
PubChem CID 2734062
ChEBI CHEBI:18662
IUPAC Name (2R)-2-(chloromethyl)oxirane
SMILES ClC[C@H]1CO1

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
UN Number 2023
Formula Weight 92.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-Epichlorohydrin
EINECSNumber (2)-0275
RTECSNumber RR0427000
TSCA No