missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-DTBM-SEGPHOS(regR) 99.0+%, TCI America™
$117.43 - $320.62
Chemical Identifiers
| CAS | 566940-03-2 |
|---|---|
| Molecular Formula | C74H100O8P2 |
| Molecular Weight (g/mol) | 1179.554 |
| MDL Number | MFCD09753003 |
| InChI Key | ZNORAFJUESSLTM-UHFFFAOYSA-N |
| Synonym | (R)-(-)-5,5′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (R)-(-)-4,4′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene) |
| PubChem CID | 11194192 |
| IUPAC Name | [4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane |
| SMILES | CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C |
Chemical Identifiers
| 566940-03-2 | |
| 1179.554 | |
| ZNORAFJUESSLTM-UHFFFAOYSA-N | |
| 11194192 | |
| CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C |
| C74H100O8P2 | |
| MFCD09753003 | |
| (R)-(-)-5,5′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (R)-(-)-4,4′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene) | |
| [4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane |
Specifications
| 566940-03-2 | |
| White | |
| MFCD09753003 | |
| (R)-(-)-5,5′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (R)-(-)-4,4′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene) | |
| CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C | |
| 1179.554 | |
| 1179.55 | |
| Crystalline Powder |
| 128°C | |
| C74H100O8P2 | |
| 200 mg | |
| ZNORAFJUESSLTM-UHFFFAOYSA-N | |
| [4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane | |
| 11194192 | |
| ≥99.0% (HPLC) | |
| (R)-(-)-DTBM-SEGPHOS(regR) |
Safety and Handling
TSCA : No