accessibility menu, dialog, popup

UserName

CAS RN 566940-03-2

CH3CH3H3COCH3CH3CH3H3CPOOOOPCH3H3CCH3OCH3CH3CH3H3CCH3CH3H3COH3CH3CCH3H3CH3CH3CCH3OH3CCH3H3CCH3(?)

CAS RN 566940-03-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results