missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-DBD-Py-NCS 98.0+%, TCI America™

Catalog No. a5568100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5568100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5568100MG Supplier TCI America Supplier No. A5568100MG

[=(R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole],[for HPLC Labeling]

Chemical Identifiers

CAS 163927-31-9
Molecular Formula C13H15N5O3S2
Molecular Weight (g/mol) 353.415
MDL Number MFCD00671493
InChI Key YMXYIIXTKUQWPC-SECBINFHSA-N
Synonym (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole
PubChem CID 16217521
IUPAC Name 7-[(3R)-3-isothiocyanatopyrrolidin-1-yl]-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)N3CCC(C3)N=C=S

Specifications

Quantity 100 mg
Formula Weight 353.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-DBD-Py-NCS [=(R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl
TSCA No
Recommended Storage Refrigerator