CAS RN 163927-31-9
IUPAC Name:
7-(3-isothiocyanatopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Synonyms:
7-(3-isothiocyanatopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Molecular Weight (g/mol):
353.42
Molecular Formula:
C13H15N5O3S2
InChi Key:
YMXYIIXTKUQWPC-UHFFFAOYNA-N
SMILES:
CN(C)S(=O)(=O)C1=CC=C(N2CCC(C2)N=C=S)C2=NON=C12
1
–
1
of
1
results
1
(R)-(-)-DBD-Py-NCS 98.0+%, TCI America™
CAS: 163927-31-9 Molecular Formula: C13H15N5O3S2 Molecular Weight (g/mol): 353.415 MDL Number: MFCD00671493 InChI Key: YMXYIIXTKUQWPC-SECBINFHSA-N Synonym: (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole PubChem CID: 16217521 IUPAC Name: 7-[(3R)-3-isothiocyanatopyrrolidin-1-yl]-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide SMILES: CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)N3CCC(C3)N=C=S