missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-Carvone 99.0+%, TCI America™
Supplier: TCI America C0703100ML
Specifications
| (R)-(-)-Carvone | |
| Yellow | |
| C10H14O | |
| 100 mL | |
| ULDHMXUKGWMISQ-SECBINFHSA-N | |
| (5R)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one | |
| 439570 | |
| 150.22 | |
| Liquid |
| 6485-40-1 | |
| 231°C | |
| MFCD00001578 | |
| --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone | |
| CC(=C)[C@@H]1CC=C(C)C(=O)C1 | |
| 150.22 | |
| CHEBI:15400 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 6485-40-1 | |
| 150.22 | |
| ULDHMXUKGWMISQ-SECBINFHSA-N | |
| 439570 | |
| (5R)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one |
| C10H14O | |
| MFCD00001578 | |
| --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone | |
| CHEBI:15400 | |
| CC(=C)[C@@H]1CC=C(C)C(=O)C1 |
Safety and Handling
EINECSNumber : (3)-2402
RTECSNumber : OS8650000
TSCA : Yes