missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium Tetrafluoroborate 97.0+%, TCI America™
$216.24 - $747.49
Chemical Identifiers
| CAS | 862095-77-0 |
|---|---|
| Molecular Formula | C19H17BF7N3 |
| Molecular Weight (g/mol) | 431.165 |
| InChI Key | UDDOGJQUYCEOBY-UNTBIKODSA-N |
| PubChem CID | 11154665 |
| IUPAC Name | (5R)-5-benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-4-ium;tetrafluoroborate |
| SMILES | [B-](F)(F)(F)F.C1CC2=NN(C=[N+]2C1CC3=CC=CC=C3)C4=CC=C(C=C4)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3592200MG
|
TCI America
B3592200MG |
200 mg |
Each for $216.24
|
|
|||||
|
B35921G
|
TCI America
B35921G |
1 g |
Each for $747.49
|
|
|||||
Chemical Identifiers
| 862095-77-0 | |
| 431.165 | |
| 11154665 | |
| [B-](F)(F)(F)F.C1CC2=NN(C=[N+]2C1CC3=CC=CC=C3)C4=CC=C(C=C4)C(F)(F)F |
| C19H17BF7N3 | |
| UDDOGJQUYCEOBY-UNTBIKODSA-N | |
| (5R)-5-benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-4-ium;tetrafluoroborate |
Specifications
| 862095-77-0 | |
| White-Yellow | |
| 200 mg | |
| UDDOGJQUYCEOBY-UNTBIKODSA-N | |
| (5R)-5-benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-4-ium;tetrafluoroborate | |
| 11154665 | |
| ≥97.0% (N) | |
| (R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium Tetrafluorob |
| 203°C | |
| C19H17BF7N3 | |
| 1759 | |
| [B-](F)(F)(F)F.C1CC2=NN(C=[N+]2C1CC3=CC=CC=C3)C4=CC=C(C=C4)C(F)(F)F | |
| 431.165 | |
| 431.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No