missing translation for 'onlineSavingsMsg'
Learn More

(R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium Tetrafluoroborate 97.0+%, TCI America™

Catalog No. B35921G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35921G 1 g
B3592200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35921G Supplier TCI America Supplier No. B35921G

Chemical Identifiers

CAS 862095-77-0
Molecular Formula C19H17BF7N3
Molecular Weight (g/mol) 431.165
InChI Key UDDOGJQUYCEOBY-UNTBIKODSA-N
PubChem CID 11154665
IUPAC Name (5R)-5-benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-4-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1CC2=NN(C=[N+]2C1CC3=CC=CC=C3)C4=CC=C(C=C4)C(F)(F)F

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 431.16
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium Tetrafluorob