missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl Chloride 99.0+%, TCI America™

Catalog No. p-7132541
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1104100MG 100 mg
M11041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1104100MG Supplier TCI America Supplier No. M1104100MG

[for Determination of the optical purity of Alcohols and Amines]

Chemical Identifiers

CAS 39637-99-5
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.62
MDL Number MFCD00044400
InChI Key PAORVUMOXXAMPL-VIFPVBQESA-N
Synonym unii-81ut10uhv3, r---mtpa-cl, --alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, r---a-methoxy-a-trifluoromethylphenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, --mtpa-cl, --mosher's acid chloride
PubChem CID 3080792
IUPAC Name (2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 99°C
Quantity 100 mg
UN Number 3265
Formula Weight 252.62
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl Chloride [for Determination of the optical