missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl Chloride, TCI America™

Catalog No. M22155G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22155G Supplier TCI America Supplier No. M22155G

(ca. 18% in Dichloromethane, ca. 1.0mol/L) [for Determination of the optical purity of Alcohols and Amines]

Chemical Identifiers

CAS 39637-99-5
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.62
MDL Number MFCD00044400
InChI Key PAORVUMOXXAMPL-VIFPVBQESA-N
Synonym unii-81ut10uhv3, r---mtpa-cl, --alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, r---a-methoxy-a-trifluoromethylphenylacetyl chloride, r---alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, --mtpa-cl, --mosher's acid chloride
PubChem CID 3080792
IUPAC Name (2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 252.62
Physical Form Liquid
Chemical Name or Material (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl Chloride (ca. 18% in Dichloromethane, ca.
TSCA No
Recommended Storage Refrigerator