missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™

Catalog No. M08311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08311G 1 g
M08315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08311G Supplier TCI America Supplier No. M08311G

Chemical Identifiers

CAS 20445-31-2
Molecular Formula C10H9F3O3
Molecular Weight (g/mol) 234.17
MDL Number MFCD00004184
InChI Key JJYKJUXBWFATTE-UHFFFAOYNA-N
Synonym +-mtpa, unii-27o5l9t1wm, r-moshers acid, +-mosher's acid, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-alpha-methoxy-alpha-trifluoromethylphenylacetic acid, r-mtpa, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-a-methoxy-a-trifluoromethylphenylacetic acid, r-3,3,3-trifluoro-2-methoxy-2-phenyl-propionic acid
PubChem CID 2723917
IUPAC Name 3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
SMILES COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Color White
Quantity 1 g
Formula Weight 234.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic Acid [Optical Resolving]