missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-alpha-Lipoic Acid 98.0+%, TCI America™
Supplier: TCI America L020725G
Specifications
| (R)-alpha-Lipoic Acid | |
| 49°C | |
| C8H14O2S2 | |
| 25 g | |
| AGBQKNBQESQNJD-SSDOTTSWSA-N | |
| 5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid | |
| 6112 | |
| 206.32 | |
| Crystalline Powder |
| 1200-22-2 | |
| Yellow | |
| MFCD01631142 | |
| (R)-Thioctic Acid, (R)-1,2-Dithiolane-3-valeric Acid | |
| OC(=O)CCCC[C@@H]1CCSS1 | |
| 206.32 | |
| CHEBI:30314 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1200-22-2 | |
| 206.32 | |
| AGBQKNBQESQNJD-SSDOTTSWSA-N | |
| 6112 | |
| 5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid |
| C8H14O2S2 | |
| MFCD01631142 | |
| (R)-Thioctic Acid, (R)-1,2-Dithiolane-3-valeric Acid | |
| CHEBI:30314 | |
| OC(=O)CCCC[C@@H]1CCSS1 |
Safety and Handling
EINECSNumber : (5)-0535
TSCA : No
Recommended Storage : Refrigerator