accessibility menu, dialog, popup

UserName

HOO(R)SS

CAS RN 1200-22-2

IUPAC Name: 5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid
Synonyms: lipoic acid pleomix α pleomix-α DL-α-lipoic acid pyruvate oxidation factor tioctidasi espa-lipon duralipon lipoic acid (substance) trommsdorgg brand of thioctic acid
Molecular Weight (g/mol): 206.32
Molecular Formula: C8H14O2S2
InChi Key: AGBQKNBQESQNJD-SSDOTTSWSA-N
SMILES: OC(=O)CCCC[C@@H]1CCSS1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results