missing translation for 'onlineSavingsMsg'
Learn More

(R)-alpha-Lipoic Acid 98.0+%, TCI America™

Supplier:  TCI America L0207100G

 View more versions of this product

Catalog No. L0207100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(R)-alpha-Lipoic Acid
49°C
C8H14O2S2
100 g
AGBQKNBQESQNJD-SSDOTTSWSA-N
5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid
6112
206.32
Crystalline Powder
1200-22-2
Yellow
MFCD01631142
(R)-Thioctic Acid, (R)-1,2-Dithiolane-3-valeric Acid
OC(=O)CCCC[C@@H]1CCSS1
206.32
CHEBI:30314
≥98.0% (HPLC,T)

Chemical Identifiers

1200-22-2
206.32
AGBQKNBQESQNJD-SSDOTTSWSA-N
6112
5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid
C8H14O2S2
MFCD01631142
(R)-Thioctic Acid, (R)-1,2-Dithiolane-3-valeric Acid
CHEBI:30314
OC(=O)CCCC[C@@H]1CCSS1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0535

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions