missing translation for 'onlineSavingsMsg'
Learn More

(R)-alpha-Lipoic Acid 98.0+%, TCI America™

Catalog No. L02075G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02075G 5 g
L020725G 25 g
L0207100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L02075G Supplier TCI America Supplier No. L02075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1200-22-2
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD01631142
InChI Key AGBQKNBQESQNJD-SSDOTTSWSA-N
Synonym (R)-Thioctic Acid, (R)-1,2-Dithiolane-3-valeric Acid
PubChem CID 6112
ChEBI CHEBI:30314
IUPAC Name 5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid
SMILES OC(=O)CCCC[C@@H]1CCSS1

Specifications

Melting Point 49°C
Color Yellow
Quantity 5 g
Formula Weight 206.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-alpha-Lipoic Acid
EINECSNumber (5)-0535
TSCA No
Recommended Storage Refrigerator