missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol 98.0+%, TCI America™

Catalog No. D23651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23651G 1 g
D23655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23651G Supplier TCI America Supplier No. D23651G

Chemical Identifiers

CAS 22348-32-9
Molecular Formula C17H19NO
Molecular Weight (g/mol) 253.345
MDL Number MFCD00077754
InChI Key OGCGXUGBDJGFFY-MRXNPFEDSA-N
Synonym r-diphenyl pyrrolidin-2-yl methanol, r-+-alpha,alpha-diphenyl-2-pyrrolidinemethanol, r-+-2-diphenylhydroxymethyl pyrrolidine, diphenyl 2r-pyrrolidin-2-ylmethanol, r-+-2-diphenylhydroxymethyl-pyrrolidin, diphenyl-2r-pyrrolidin-2-yl methanol, r-alpha,alpha-diphenyl-2-pyrrolidinemethanol, alpha,alpha-diphenyl-d-prolinol, r-+-alpha,alpha-diphenylprolinol, diphenyl 2r-pyrrolidin-2-yl methanol
PubChem CID 7045371
IUPAC Name diphenyl-[(2R)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 78°C
Color White
Quantity 1 g
Formula Weight 253.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol