missing translation for 'onlineSavingsMsg'
Learn More

(R)-5-(Hydroxymethyl)-2-pyrrolidinone 98.0+%, TCI America™

Catalog No. H12415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12415G 5 g
H12411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12415G Supplier TCI America Supplier No. H12415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66673-40-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00077791
InChI Key HOBJEFOCIRXQKH-SCSAIBSYSA-N
Synonym r---5-hydroxymethyl-2-pyrrolidinone, d-pyroglutaminol, 5r-5-hydroxymethyl pyrrolidin-2-one, r-5-hydroxymethyl pyrrolidin-2-one, r-5-hydroxymethyl-2-pyrrolidinone, r-5-hydroxymethyl-pyrrolidin-2-one, --d-pyroglutamol, r---5-hydroxymethylpyrrolidin-2-one, 5-hydroxymethyl-2-pyrrolidinone
PubChem CID 7128291
IUPAC Name (5R)-5-(hydroxymethyl)pyrrolidin-2-one
SMILES C1CC(=O)NC1CO

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 5 g
Formula Weight 115.13
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-5-(Hydroxymethyl)-2-pyrrolidinone