accessibility menu, dialog, popup

UserName

CAS RN 66673-40-3

IUPAC Name: (5R)-5-(hydroxymethyl)pyrrolidin-2-one
Synonyms: (5R)-5-(hydroxymethyl)pyrrolidin-2-one
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: HOBJEFOCIRXQKH-SCSAIBSYSA-N
SMILES: OC[C@H]1CCC(=O)N1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)