missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol 99.0+%, TCI America™

Catalog No. p-7134275
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02821G 1 g
O02825G 5 g
O028225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O02821G Supplier TCI America Supplier No. O02821G
Only null left

Chemical Identifiers

CAS 65355-14-8
Molecular Formula C20H22O2
Molecular Weight (g/mol) 294.394
MDL Number MFCD02093485
InChI Key UTXIFKBYNJRJPH-UHFFFAOYSA-N
Synonym s---5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, r-+-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, r-5,5',6,6',7,7',8,8'-octahydro 1,1'-binaphthalene-2,2'-diol, s-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthalene-2,2'-diol, 5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthalene-2,2'-diol, 5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, 5,5',6,6',7,7',8,8'-octahydro-bi-2-naphthol, r-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, s---2,2'-dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, 1-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl-5,6,7,8-tetrahydronaphthalen-2-ol
PubChem CID 3694111
IUPAC Name 1-(2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl)-5,6,7,8-tetrahydronaphthalen-2-ol
SMILES C1CCC2=C(C1)C=CC(=C2C3=C(C=CC4=C3CCCC4)O)O

Specifications

Melting Point 167°C
Color White
Quantity 1 g
Formula Weight 294.39
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-bi-2-naphthol