missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-4-Phenylthiazolidine-2-thione 98.0+%, TCI America™
Supplier: TCI America P19591G
Specifications
| (R)-4-Phenylthiazolidine-2-thione | |
| White-Yellow | |
| MFCD06658215 | |
| IEXSISKCCADMLK-QMMMGPOBSA-N | |
| (4R)-4-phenyl-1,3-thiazolidine-2-thione | |
| 11521249 | |
| ≥98.0% (GC) |
| 110199-18-3 | |
| C9H9NS2 | |
| 1 g | |
| C1C(NC(=S)S1)C2=CC=CC=C2 | |
| 195.298 | |
| 195.30 | |
| Crystalline Powder |
Chemical Identifiers
| 110199-18-3 | |
| 195.298 | |
| IEXSISKCCADMLK-QMMMGPOBSA-N | |
| (4R)-4-phenyl-1,3-thiazolidine-2-thione |
| C9H9NS2 | |
| MFCD06658215 | |
| 11521249 | |
| C1C(NC(=S)S1)C2=CC=CC=C2 |
Safety and Handling
TSCA : No