accessibility menu, dialog, popup

UserName

HNSS

CAS RN 110199-18-3

IUPAC Name: 4-phenyl-1,3-thiazolidine-2-thione
Synonyms: 4-phenyl-1,3-thiazolidine-2-thione
Molecular Weight (g/mol): 195.3
Molecular Formula: C9H9NS2
InChi Key: IEXSISKCCADMLK-UHFFFAOYNA-N
SMILES: S=C1NC(CS1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results