missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-Isopropyl-2-oxazolidinone 98.0+%, TCI America™

Catalog No. p-7131494
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05721G 1 g
I05725G 5 g
I057225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I05721G Supplier TCI America Supplier No. I05721G
Only null left

Chemical Identifiers

CAS 95530-58-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00075172
InChI Key YBUPWRYTXGAWJX-YFKPBYRVSA-N
Synonym r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
PubChem CID 641505
IUPAC Name (4R)-4-(propan-2-yl)-1,3-oxazolidin-2-one
SMILES CC(C)[C@@H]1COC(=O)N1

Specifications

Melting Point 72°C
Color White
Quantity 1 g
Formula Weight 129.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-4-Isopropyl-2-oxazolidinone