missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone, 97%

Catalog No. AAH5107403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5107403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5107403 Supplier Thermo Scientific Chemicals Supplier No. H5107403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 173604-33-6 | C16H15NO2 | 253.301 g/mol

(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible w

Chemical Identifiers

CAS 173604-33-6
Molecular Formula C16H15NO2
Molecular Weight (g/mol) 253.301
MDL Number MFCD01863565
InChI Key QEOCTJMBYZNEJH-AWEZNQCLSA-N
Synonym r-4-benzhydryloxazolidin-2-one, r-+-4-diphenylmethyl-2-oxazolidinone, r-4-methyldiphenyl-2-oxazolidinone, 4r-4-diphenylmethyl-1,3-oxazolidin-2-one, pubchem11686, 4r-4-benzhydryloxazolidine-2-one, r-4-diphenylmethyl oxazolidin-2-one, 2-oxazolidinone,4-diphenylmethyl-, 4r, 2-oxazolidinone, 4-diphenylmethyl-, 4r, r-+-4-diphenylmethyl-2-oxazolidin
PubChem CID 9837924
IUPAC Name (4R)-4-benzhydryl-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 253.3
Percent Purity 98%
Chemical Name or Material (R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only