missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane 98.0+%, TCI America™
Supplier: TCI America C226525G
Specifications
| (R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane | |
| Colorless | |
| C6H11ClO2 | |
| 25 g | |
| r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, r-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, 4r, r-3-chloro-1,2-propanediol acetonide, pubchem14074, ksc911s2r | |
| CC1(C)OCC(CCl)O1 | |
| 150.60 | |
| 150.60 | |
| Liquid |
| 57044-24-3 | |
| 63°C | |
| MFCD00273371 | |
| 1992 | |
| BNPOTXLWPZOESZ-UHFFFAOYNA-N | |
| 4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane | |
| 2734443 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 57044-24-3 | |
| 150.60 | |
| BNPOTXLWPZOESZ-UHFFFAOYNA-N | |
| 2734443 | |
| CC1(C)OCC(CCl)O1 |
| C6H11ClO2 | |
| MFCD00273371 | |
| r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, r-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, 4r, r-3-chloro-1,2-propanediol acetonide, pubchem14074, ksc911s2r | |
| 4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane |
Safety and Handling
TSCA : No