missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane 98.0+%, TCI America™

Catalog No. p-7125373
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22655G 5 g
C226525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22655G Supplier TCI America Supplier No. C22655G

Chemical Identifiers

CAS 57044-24-3
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00273371
InChI Key BNPOTXLWPZOESZ-UHFFFAOYNA-N
Synonym r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, r-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, 4r, r-3-chloro-1,2-propanediol acetonide, pubchem14074, ksc911s2r
PubChem CID 2734443
IUPAC Name 4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
SMILES CC1(C)OCC(CCl)O1

Specifications

Color Colorless
Boiling Point 63°C
Quantity 5 g
UN Number 1992
Formula Weight 150.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane