missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 98%

Catalog No. AAH62954MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62954MD 250 mg
AAH6295403 1 g
AAH6295406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62954MD Supplier Thermo Scientific Chemicals Supplier No. H62954MD

CAS: 114360-56-4 | C24H28N2O6 | 440.496 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114360-56-4
Molecular Formula C24H28N2O6
Molecular Weight (g/mol) 440.496
MDL Number MFCD00798647
InChI Key LIWKOFAHRLBNMG-HXUWFJFHSA-N
Synonym fmoc-d-dab boc-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, n-a-fmoc-n-g-boc-d-2,4-diaminobutyric acid, 2r-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-t-butyloxycarbonyl-d-2,4-diaminobutyric acid, ambotzfaa1317, fmoc-4-boc-d-2,4-diaminobutyric acid, n-, a-fmoc-n-, a-boc-d-2,4-diaminobutyricacid, n-a-fmoc-n-gamma-boc-d-2,4-diaminobutyric acid, 2r-4-boc-amino-2-fmoc-amino-butanoic acid
PubChem CID 7021129
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Color White
Quantity 250 mg
Formula Weight 440.49
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only