missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-Benzyloxy-1,2-butanediol 97.0+%, TCI America™

Catalog No. B2899200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2899200MG 200 mg
B28991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2899200MG Supplier TCI America Supplier No. B2899200MG
Only null left

Chemical Identifiers

CAS 86990-91-2
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
InChI Key TVRPDIKPMQUOSL-LLVKDONJSA-N
PubChem CID 10910454
IUPAC Name (2R)-4-phenylmethoxybutane-1,2-diol
SMILES C1=CC=C(C=C1)COCCC(CO)O

Specifications

Color Yellow
Boiling Point 168°C
Quantity 200 mg
Formula Weight 196.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-4-Benzyloxy-1,2-butanediol