missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-4-Benzyl-2-oxazolidinone 98.0+%, TCI America™
$127.21 - $406.31
Chemical Identifiers
| CAS | 102029-44-7 |
|---|---|
| Molecular Formula | C10H11NO2 |
| Molecular Weight (g/mol) | 177.20 |
| MDL Number | MFCD00010846 |
| InChI Key | OJOFMLDBXPDXLQ-SECBINFHSA-N |
| Synonym | r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one # |
| PubChem CID | 2734969 |
| IUPAC Name | (4R)-4-benzyl-1,3-oxazolidin-2-one |
| SMILES | O=C1N[C@H](CC2=CC=CC=C2)CO1 |
Chemical Identifiers
| 102029-44-7 | |
| 177.20 | |
| OJOFMLDBXPDXLQ-SECBINFHSA-N | |
| 2734969 | |
| O=C1N[C@H](CC2=CC=CC=C2)CO1 |
| C10H11NO2 | |
| MFCD00010846 | |
| r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one # | |
| (4R)-4-benzyl-1,3-oxazolidin-2-one |
Specifications
| 87°C | |
| White-Yellow | |
| MFCD00010846 | |
| OJOFMLDBXPDXLQ-SECBINFHSA-N | |
| (4R)-4-benzyl-1,3-oxazolidin-2-one | |
| 177.20 | |
| 177.20 | |
| Crystalline Powder |
| 102029-44-7 | |
| C10H11NO2 | |
| r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one # | |
| O=C1N[C@H](CC2=CC=CC=C2)CO1 | |
| 5 g | |
| 2734969 | |
| ≥98.0% (GC) | |
| (R)-4-Benzyl-2-oxazolidinone |
Safety and Handling
TSCA : No