accessibility menu, dialog, popup

UserName

ONH(R)O

CAS RN 102029-44-7

IUPAC Name: (4R)-4-benzyl-1,3-oxazolidin-2-one
Synonyms: (4R)-4-benzyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: OJOFMLDBXPDXLQ-SECBINFHSA-N
SMILES: O=C1N[C@H](CC2=CC=CC=C2)CO1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)