missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-Benzyl-2-oxazolidinone, 99%

CAS: 102029-44-7 | C10H11NO2 | 177.20 g/mol

Supplier:  Thermo Scientific Chemicals A1677014

 View more versions of this product

Catalog No. AAA1677014


Only null left

Description

Description

(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

Solubility
Insoluble in water. Soluble in Chloroform.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

86°C to 89°C
102029-44-7
MFCD00010846
r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one #
OJOFMLDBXPDXLQ-SECBINFHSA-N
(4R)-4-benzyl-1,3-oxazolidin-2-one
177.20
+64° (c=1 in Chloroform)
Hygroscopic
(R)-(+)-4-Benzyl-2-oxazolidinone
C10H11NO2
4782551
Insoluble in water. Soluble in Chloroform.
O=C1N[C@H](CC2=CC=CC=C2)CO1
25 g
2734969
177.2
99%

Chemical Identifiers

102029-44-7
177.20
OJOFMLDBXPDXLQ-SECBINFHSA-N
2734969
O=C1N[C@H](CC2=CC=CC=C2)CO1
C10H11NO2
MFCD00010846
r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one #
(4R)-4-benzyl-1,3-oxazolidin-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only