missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-Benzyl-2-oxazolidinone, 99%

Catalog No. p-7023336
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1677003 1 g
AAA1677006 5 g
AAA1677014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1677003 Supplier Thermo Scientific Chemicals Supplier No. A1677003
Only null left

CAS: 102029-44-7 | C10H11NO2 | 177.20 g/mol

(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

Solubility
Insoluble in water. Soluble in Chloroform.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 102029-44-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00010846
InChI Key OJOFMLDBXPDXLQ-SECBINFHSA-N
Synonym r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one #
PubChem CID 2734969
IUPAC Name (4R)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@H](CC2=CC=CC=C2)CO1

Specifications

Melting Point 86°C to 89°C
Beilstein 4782551
Solubility Information Insoluble in water. Soluble in Chloroform.
Quantity 1 g
Optical Rotation +64° (c=1 in Chloroform)
Formula Weight 177.2
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material (R)-(+)-4-Benzyl-2-oxazolidinone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only