missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-Amino-2-methyl-1-butanol 98.0+%, TCI America™

Catalog No. A15761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15761G 1 g
A15765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15761G Supplier TCI America Supplier No. A15761G
Only null left

Chemical Identifiers

CAS 88390-32-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD03093614
InChI Key DUAXLVGFFDFSAG-RXMQYKEDSA-N
PubChem CID 11457670
IUPAC Name (2R)-4-amino-2-methylbutan-1-ol
SMILES CC(CCN)CO

Specifications

Color Yellow
Boiling Point 80°C
Quantity 1 g
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-4-Amino-2-methyl-1-butanol
EINECSNumber (2)-3927
TSCA No