accessibility menu, dialog, popup

UserName

CH3H2NOH

CAS RN 88390-32-3

IUPAC Name: 4-amino-2-methylbutan-1-ol
Synonyms: 4-amino-2-methylbutan-1-ol
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: DUAXLVGFFDFSAG-UHFFFAOYNA-N
SMILES: CC(CO)CCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results