missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane 94.0+%, TCI America™

Catalog No. H11881G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H11881G Supplier TCI America Supplier No. H11881G

Chemical Identifiers

CAS 70005-89-9
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD02682966
InChI Key YYEZYENJAMOWHW-ZCFIWIBFSA-N
Synonym (R)-2-(2,2-Dimethyl-1,3-dioxolan-4-yl)ethanol
PubChem CID 11480522
IUPAC Name 2-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethanol
SMILES CC1(OCC(O1)CCO)C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 146.19
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane
EINECSNumber (5)-0502
TSCA No