accessibility menu, dialog, popup

UserName

H3COOHOCH3

CAS RN 70005-89-9

IUPAC Name: 2-(2,2-dimethyl-1,3-dioxolan-4-yl)ethan-1-ol
Synonyms: 2-(2,2-dimethyl-1,3-dioxolan-4-yl)ethanol
Molecular Weight (g/mol): 146.19
Molecular Formula: C7H14O3
InChi Key: YYEZYENJAMOWHW-UHFFFAOYNA-N
SMILES: CC1(C)OCC(CCO)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results