missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Fmoc-amino)-3-phenylpropionic acid, 95%

Catalog No. AAH52074MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52074MD 250 mg
AAH5207403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52074MD Supplier Thermo Scientific Chemicals Supplier No. H52074MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 220498-02-2 | C24H21NO4 | 387.435 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 220498-02-2
Molecular Formula C24H21NO4
Molecular Weight (g/mol) 387.435
MDL Number MFCD01311783
InChI Key PTSLRPMRTOVHAB-JOCHJYFZSA-N
Synonym r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-3-amino-3-phenylpropionic acid, fmoc-l-beta-phenylalanine, fmoc-beta-phe-oh, r-n-fmoc-3-amino-3-phenylpropanoic acid, r-3-fmoc-amino-3-phenylpropionic acid, fmoc-r-phenylalanine, 3r-3-9h-fluoren-9-ylmethoxycarbonylamino-3-phenylpropanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid, fmoc-d-?-phe-oh
PubChem CID 2734302
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 250 mg
Beilstein 7982022
Formula Weight 387.43
Percent Purity 95%
Chemical Name or Material (R)-3-(Fmoc-amino)-3-phenylpropionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only