missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 98%

CAS: 517905-93-0 | C24H20N2O6 | 432.43 g/mol

Supplier:  Thermo Scientific Chemicals H5203706

 View more versions of this product

Catalog No. AAH5203706


Only null left
Explore available promotions

Description

Description

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid
170°C to 172°C
MFCD03428045
r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-2-nitrophenyl propanoic acid, fmoc-r-3-amino-3-2-nitrophenyl-propionic acid, fmoc-d-beta-phe 2-no2-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-nitrophenyl propanoic acid, pubchem24026, fmoc-beta-phe 2-no2-oh, fmoc-d-?-phe 2-no2-oh, n-fmoc-2-nitro-l-beta-phenylalanine, fmoc-r-3-amino-3-2-nitro-phenyl-propionic acid, r-3-fmoc-amino-3-2-nitrophenyl propionic acid
DRESTDPDCGPKNI-UHFFFAOYNA-N
OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O
432.43
432.43
517905-93-0
C24H20N2O6
5 g
Slightly soluble in water.
−182° (c=1 in DMF)
3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
2761743
98%

Chemical Identifiers

517905-93-0
432.43
DRESTDPDCGPKNI-UHFFFAOYNA-N
2761743
OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O
C24H20N2O6
MFCD03428045
r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-2-nitrophenyl propanoic acid, fmoc-r-3-amino-3-2-nitrophenyl-propionic acid, fmoc-d-beta-phe 2-no2-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-nitrophenyl propanoic acid, pubchem24026, fmoc-beta-phe 2-no2-oh, fmoc-d-?-phe 2-no2-oh, n-fmoc-2-nitro-l-beta-phenylalanine, fmoc-r-3-amino-3-2-nitro-phenyl-propionic acid, r-3-fmoc-amino-3-2-nitrophenyl propionic acid
3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only