missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 98%

Catalog No. AAH52037MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52037MD 250 mg
AAH5203703 1 g
AAH5203706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52037MD Supplier Thermo Scientific Chemicals Supplier No. H52037MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 517905-93-0 | C24H20N2O6 | 432.43 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 517905-93-0
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD03428045
InChI Key DRESTDPDCGPKNI-UHFFFAOYNA-N
Synonym r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-2-nitrophenyl propanoic acid, fmoc-r-3-amino-3-2-nitrophenyl-propionic acid, fmoc-d-beta-phe 2-no2-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-nitrophenyl propanoic acid, pubchem24026, fmoc-beta-phe 2-no2-oh, fmoc-d-?-phe 2-no2-oh, n-fmoc-2-nitro-l-beta-phenylalanine, fmoc-r-3-amino-3-2-nitro-phenyl-propionic acid, r-3-fmoc-amino-3-2-nitrophenyl propionic acid
PubChem CID 2761743
IUPAC Name 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
SMILES OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 170°C to 172°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Optical Rotation −182° (c=1 in DMF)
Formula Weight 432.43
Percent Purity 98%
Chemical Name or Material (R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only